missing translation for 'onlineSavingsMsg'
Learn More

Cetirizine.2HCl, Lipomed

Product Code. 30460112
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30460112 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460112 Supplier Lipomed Supplier No. LPMCET1871HC50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Cetirizine.2HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cetirizine Dihydrochloride
CAS 83881-52-1
Analyte or Component Names Cetirizine Dihydrochloride
Shipping Condition Room Temperature
Molecular Formula C21 H25 Cl N2 O3 . 2 Cl H
Synonym Cetirizine Dihydrochloride, Acetic acid, [2-[4-[(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]-, dihydrochloride (9 CI), Alercet, Alerid, Alerlisin, Cesta, Cetirizine hydrochloride, Cetrine, Cetrizet, Cistamine, Formistin, Histazine, Nosemin, P 071, Reactine, Riztec, Ryzen, Sancotec, Selitex, Triz, UCB-P 071, Virlix, Zeran, Zirtec, Zirtek, Zirtin, Zyrlex, Zyrtec, Zyrzine, Cetiriz, Cetirizine dihydrochloride, Cetrak, Epirizine, Histazine-1, Tomazine
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Cl.Cl.OC(=O)COCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3
IUPAC Name 2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetic acid;dihydrochloride
Industry Type Pharmaceutical
Molecular Weight (g/mol) 461.81
Quantity 50 mg
Formula Weight 460.1087 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.