missing translation for 'onlineSavingsMsg'
Learn More

Cesium Acetate, 99.9%, Optical Grade, MP Biomedicals™

Product Code. 11463250 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463250 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11463250 Supplier MP Biomedicals Supplier No. 0210131925

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 3396-11-0
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.9%
Molecular Formula C2H3CsO2
Synonym cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ion
InChI Key ZOAIGCHJWKDIPJ-UHFFFAOYSA-M
SMILES CC(=O)[O-].[Cs+]
IUPAC Name cesium;acetate
Molecular Weight (g/mol) 191.949
PubChem CID 5152919
Formula Weight 191.9
Grade Optical
Chemical Name or Material Cesium Acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.