Learn More
Thermo Scientific™ Ceramic Homogenizers for QuEChERS Kits
Product Code. 17349822 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
15 mL Tubes
2 mL Tubes
50 mL Tubes
Unit Size:
Pack of 100
Pack of 200
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17349822 50 mL Tubes Pack of 100
17329822 15 mL Tubes Pack of 100
17339822 2 mL Tubes Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17349822 Supplier Thermo Scientific™ Supplier No. 60106CH50
Save Up to 60% on Contract Testing Essentials
Use promo code 28025
View all promotions
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
Use Thermo Scientific™ Ceramic Homogenizers to grind and homogenize samples for your QuEChERS sample preparation workflow.
Use Thermo Scientific™ Ceramic Homogenizers to help accelerate the homogenization process and increase recovery and reproducibility during your QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) sample preparation workflow. These homognizers can be used with all of our QuEChERS Extraction and Clean-Up kits to supplement Original 2003 Method, AOAC 2007.01 Method, EN 15662 Method, and NY/T 1380-2007 Method procedures.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 50 mL Tubes
Type Ceramic Homogenizer
Quantity 100/Pk.