missing translation for 'onlineSavingsMsg'
Learn More

Cepharanthine, Thermo Scientific™

Product Code. 17730639 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17730639 250 mg 250mg
17720639 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17730639 Supplier Thermo Scientific Acros Supplier No. 468912500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Chemical Name or Material Cepharanthine
CAS 481-49-2
Quantity 250 mg
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C37 H38 N2 O6
Physical Form Crystalline Powder
Color White to Light-yellow to Beige
Purity ≥94%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.