Specifications
| Chemical Name or Material | Cepharanthine |
| CAS | 481-49-2 |
| Quantity | 1 g |
| Shipping Condition | No special requirements |
| Product Line | Life science |
| Molecular Formula | C37 H38 N2 O6 |
| Physical Form | Crystalline Powder |
| Color | White to Light-yellow to Beige |
| Purity | ≥94% |