missing translation for 'onlineSavingsMsg'
Learn More

Cephapirin Benzathine, TRC

Product Code. 30134879
Change view
Click to view available options
Quantity:
10 mg
1 g
Unit Size:
10mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134879 1 g 1g
17934475 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134879 Supplier Toronto Research Chemicals Supplier No. TRCC2615051G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97468-37-6
Molecular Formula 2 C17 H17 N3 O6 S2 . C16 H20 N2
Molecular Weight (g/mol) 1087.27
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[(acetyloxy)methyl]-8-oxo-7-[[(4-pyridinylthio)acetyl]amino]-, (6R,7R)-, compd. with N,N'-bis(phenylmethyl)-1,2-ethanediamine (2:1), 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[(acetyloxy)methyl]-8-oxo-7-[[(4-pyridinylthio)acetyl]amino]-, (6R-trans)-, compd. with N,N'-bis(phenylmethyl)-1,2-ethanediamine (2:1), 1,2-Ethanediamine, N,N'-bis(phenylmethyl)-, bis[(6R,7R)-3-[(acetyloxy)methyl]-8-oxo-7-[[(4-pyridinylthio)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate] (9CI), Cephapirin benzathine, Metricure, Cefapirin benzathine
IUPAC Name (6R,7R)-3-(acetyloxymethyl)-8-oxo-7-[(2-pyridin-4-ylsulfanylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;N,N'-dibenzylethane-1,2-diamine
SMILES CC(=O)OCC1=C(N2[C@H](SC1)[C@H](NC(=O)CSc3ccncc3)C2=O)C(=O)O.CC(=O)OCC4=C(N5[C@H](SC4)[C@H](NC(=O)CSc6ccncc6)C5=O)C(=O)O.C(CNCc7ccccc7)NCc8ccccc8

Specifications

Quantity 1 g
InChI Formula InChI=1S/2C17H17N3O6S2.C16H20N2/c2*1-9(21)26-6-10-7-28-16-13(15(23)20(16)14(10)17(24)25)19-12(22)8-27-11-2-4-18-5-3-11;1-3-7-15(8-4-1)13-17-11-12-18-14-16-9-5-2-6-10-16/h2*2-5,13,16H,6-8H2,1H3,(H,19,22)(H,24,25);1-10,17-18H,11-14H2/t2*13-,16-;/m11./s1
Formula Weight 1086.2744
Chemical Name or Material Cephapirin Benzathine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.