Learn More
Labconco CentriZap Pro Strobe Light
Product Code. 30533440 Shop All Labconco Corporation Products
Change view
Click to view available options
Rotor System:
CentriVap Pro
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Rotor System Unit Size
30533440 CentriVap Pro Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30533440 Supplier Labconco Supplier No. 8547200
  • ONLINE EXCLUSIVE PRICE
    84.25 EUR
    78.16 EUR / Pack of 1
    Save 6.09 EUR
    7% Off
 Restricted Item
Estimated Shipment: 25-01-2027
The CentriZap™ Strobe Light enables users to view samples as if they were stationary while rotor is spinning, eliminating the need to pause a cycle.
  • For use with Labconco CentriVap Pro Vacuum Concentrators.
  • Includes mounting holster and hardware for attachment to the right side of the CentriVap Pro Vacuum Concentrator.
  • Shipping weight 2.0 lbs. (0.9 kg)
Specifications
Type Strobe Light
Rotor System CentriVap Pro