Learn More
TPP™ Centrifuge Tube with Aerosol Tight Cap
Click to view available options
Capacity (Metric):
50 mL
Unit Size:
Pack of 320
Description
Keep your samples secure and stable, these centrifuge tubes have a gas and aerosol tight cap that cannot be over tightened. The caps also feature a white writing circle for easy labelling and this allows for quick location of the sample in a rack.
- White writing circle on the cap
- Caps cannot be over tightened
- Gas and aerosol tight cap
- Graduated tubes
- Volume accuracy to ±2%
Specifications
Specifications
| Closure Color | Yellow |
| Capacity (Metric) | 50 mL |
| Material | Polypropylene |
| For Use With | Centrifugation |
| No. per Pack | 1 Ea. |
| Packaging | Sterile sub-packs of 20 |
| Shape | Conical |
| Length (Metric) | 115 mm |
| Bottom Shape | Conical |
| Writing Area | Yes |
| Show More |