Learn More
Eppendorf™ Centrifuge Adapters For Rotors for 5810 and 5810R
Product Code. 10326802 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
1.2 to 5 mL
1.5 to 2 mL
15 mL
18 to 30 mL
180 to 250 mL
2.6 to 7 mL
3 to 15 mL
30 to 50 mL
50 mL
50 to 75 mL
7 to 18 mL
80 to 120 mL
Unit Size:
Pack of 2
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
10326802 180 to 250 mL Pack of 2
10293172 1.2 to 5 mL Pack of 2
11518623 1.5 to 2 mL Pack of 2
10554332 15 mL Pack of 2
10134961 2.6 to 7 mL Pack of 2
11528623 3 to 15 mL Pack of 2
11548623 30 to 50 mL Pack of 2
10776695 18 to 30 mL Pack of 2
10102082 50 to 75 mL Pack of 2
10106771 50 mL Pack of 2
10253222 7 to 18 mL Pack of 2
11558623 80 to 120 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 10326802 Supplier Eppendorf™ Supplier No. 5810770009
Estimated Shipment: 09-10-2026
Available in a variety of configurations to suit a wide range of applications
  • Adaptors for Eppendorf™ Centrifuges 5810 and 5810R.
  • Compatible with conical or flat tubes
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Tube Type Flat
Capacity 180 to 250 mL
For Use With (Equipment) Rotors 5810 and 5810R Centrifuge
Bore Size (Metric) 62 mm
No. of Places 4
No. per Pack 2 Pack