Description
- Big Volume Harvesting Package
- With Rotor S-4xUniversal, incl. universal buckets and 1 L adapters (4x), Eppendorf 1 L bottles (4x)
- Refrigerated
- Aerosol-tight Eppendorf QuickLock™ caps and lids allow fast and ergonomic lid locking
- Centrifuge lid with soft-touch lid closure
- Quiet operation to improve your work environment
- Compact footprint saves valuable bench space
- Automatic rotor recognition and imbalance detection for maximum operational safety
Specifications
Specifications
| Max. RCF | 22,132 x g |
| Height (English) Exterior | 14.5 in. |
| Timer | 10 sec. to 99 hr. 59 min., with continuous run function, short-spin |
| Width (English) Exterior | 28.3 in. |
| Width (Metric) Exterior | 72 cm |
| Wattage | 1650 w |
| Certifications/Compliance | UL (120 V), CAN/CSA (120 V), CE, RoHS |
| Electrical Requirements | 230 V 50/60 Hz |
| Program Storage | 99 (5 program keys) |
| Profile (Acceleration/Braking) | 10/10 |
| Show More |