Learn More
Eppendorf™ Centrifuge 5420
BIOPHARMA
Product Code. 16372288 Shop All Eppendorf Products
Change view
Click to view available options
Electrical Requirements:
230 V, 50/60 Hz (EU)
230 V/50/60 Hz (EU)
Includes:
Aerosol-tight Rotor FA-24x2
Without rotor
Model:
Centrifuge 5420
Centrifuge 5420 (EU-IVD)
Unit Size:
1 set
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Electrical Requirements Includes Model Unit Size
16372288 230 V, 50/60 Hz (EU) Aerosol-tight Rotor FA-24x2 Centrifuge 5420 Each
16458520 230 V, 50/60 Hz (EU) Aerosol-tight Rotor FA-24x2 Centrifuge 5420 (EU-IVD) 1 set
16488520 230 V/50/60 Hz (EU) Without rotor Centrifuge 5420 (EU-IVD) Each
16408530 230 V, 50/60 Hz (EU) Without rotor Centrifuge 5420 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16372288 Supplier Eppendorf™ Supplier No. 5420000113
  • ONLINE EXCLUSIVE PRICE
    2520.00 EUR
    2344.00 EUR / Pack of 1
    Save 176.00 EUR
    6% Off
View all promotions
 Restricted Item
Estimated Shipment: 20-10-2026
The Eppendorf Centrifuge 5420 offers 24-place capacity in a small footprint and supports nucleic acid purification and gel-extraction applications.
  • Standard rotor for up to 24 x 1.5 and 2.0 mL tubes
  • Rotor option for up to 32 x 0.2 mL PCR tubes or 4 x 8-place PCR strips
  • Adapters for 0.2, 0.4 and 0.5 mL tubes as well as 0.6 mL BD Microtainer™
  • Soft-touch one-finger lid closure for ergonomic lid locking
  • Automatic lid opening at the end of the run to prevent sample warming and to allow easy access to samples
  • High quality anodized aluminum rotors and rotor lids
  • One push short-spin key for quick centrifugation - no need to hold key all the while
  • Easy to clean keypad control panel
  • Particularly small footprint to save precious space on the lab bench: 24 × 35 × 22 cm / 9.4 × 13.8 × 8.7 in (W × D × H)
  • Max. speed: 21,300 × g (15,060 rpm)
  • TRUSTED_SUSTAINABILITY
    Specifications
    Max. Speed 15,060 rpm
    Max. RCF 21,300 x g
    Height (English) Exterior 8.7 in.
    Height (Metric) Exterior 22 cm
    Noise Level <56 dB(A) with Rotor FA-24 x 2
    Timer 10 sec. to 9:59 hrs., with continuous run function
    Width (English) Exterior 9.4 in.
    Width (Metric) Exterior 24 cm
    Certifications/Compliance UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers
    Includes Aerosol-tight Rotor FA-24x2
    Safety Features Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, certified aerosol-tight rotor lids
    Ventilated Yes
    Electrical Requirements 230 V, 50/60 Hz (EU)
    Controller Type Keypad
    Depth (English) 13.8 in.
    Depth (Metric) 35 cm
    Model Centrifuge 5420
    Wattage 290 W
    Refrigerated No
    Standards EN ISO 12100, EN 50581, EN 61010-1, EN 61010-2-020, IEC 61010-1, IEC 61010-2-020, EN 61326-1, EN 55011, EN 378-1, EN 378-2, EN 50581 (refer to manual for more details)
    Frequency 50/60 Hz
    Voltage 230 V
    Product Type Microcentrifuge
    Display Type Large, brightly lit LCD