missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biodesign™ Cellulose Dialysis Tubing Sampler Pack
Seamless, thin, transparent dialysis tubing manufactured from the highest purity cellulose
Brand: Biodesign™ D000
Additional Details : Weight : 0.50000kg
Description
Seamless, thin, transparent dialysis tubing manufactured from the highest purity cellulose- Just wet in distilled water and use
- Available in 30.5m rolls or convenient 305mm strips
- Specially purified for life science research
- Supplied in an airtight, moisture-proof container for maximum shelf life
- Available in 14,000Da, 8,000Da, and 3,500Da molecular weight cut offs
Specifications
Dialysis Tubing BioDesign Sampler Pack | |
Cellulose | |
1.91, 3.57, 6.45 mL/cm | |
1 Each |
14 kDa | |
15.5, 21.3, 28.7 mm | |
Transparent | |
Suitable for all types of laboratory dialysis including desalting, compound separation and binding studies |