Learn More
Biodesign™ Cellulose Dialysis Tubing Sampler Pack
Seamless, thin, transparent dialysis tubing manufactured from the highest purity cellulose
284.00 EUR - 538.00 EUR
Specifications
Product Type | Dialysis Tubing BioDesign Sampler Pack |
---|---|
Material | Cellulose |
Diameter (Metric) | 15.5, 21.3, 28.7 mm |
Capacity (Metric) | 1.91, 3.57, 6.45 mL/cm |
Color | Transparent |
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
12757496
|
Biodesign™
D400 |
3.5 kDa |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12737496
|
Biodesign™
D000 |
14 kDa |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12747496
|
Biodesign™
D200 |
8 kDa |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Seamless, thin, transparent dialysis tubing manufactured from the highest purity cellulose- Just wet in distilled water and use
- Available in 30.5m rolls or convenient 305mm strips
- Specially purified for life science research
- Supplied in an airtight, moisture-proof container for maximum shelf life
- Available in 14,000Da, 8,000Da, and 3,500Da molecular weight cut offs
Specifications
Dialysis Tubing BioDesign Sampler Pack | |
15.5, 21.3, 28.7 mm | |
Transparent | |
Suitable for all types of laboratory dialysis including desalting, compound separation and binding studies |
Cellulose | |
1.91, 3.57, 6.45 mL/cm | |
1 Each |