Learn More
Cellpath System II HEX™ Cassette Lid
Product Code. 30226268
Change view
Click to view available options
Product Type:
System II HEX Cassette lid
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
30226268 System II HEX Cassette lid Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30226268 Supplier Cellpath Supplier No. EAW011408A
  • ONLINE EXCLUSIVE PRICE
    209.62 EUR
    118.25 EUR / Pack of 25
    Save 91.37 EUR
    43% Off
 Restricted Item
Estimated Shipment: 15-12-2026
For use in place of plastic lid on System II HEX tissue processing cassettes. CellPath Tissue Processing System II HEX Cassette Lid is made of stainless-steel.
Specifications
Quantity 25/Pk.
Material Stainless steel
Product Type System II HEX Cassette lid