Learn More
Nexcelom™ Cellometer Mini Bright Field Cell Counter
Product Code. 14830482
Encompass
Change view
Click to view available options
For Use With (Equipment):
Cellometer Counting Chambers
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
14830482 Cellometer Counting Chambers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 14830482 Supplier Nexcelom™ Supplier No. CELLOMETER MINI
Affordable, accurate automated cell counter for trypan blue viability
  • Small Footprint, Compatible with Most Computers
  • The Cellometer Mini utilizes bright field imaging and pattern-recognition software to quickly and accurately identify and count individual live cells and dead cells stained with Trypan Blue
  • Trypan blue cell count, concentration, diameter, and % viability are automatically calculated and reported in <10 seconds (in only 3 seconds with some computers)
  • Analysis of Clumpy & Irregular-Shaped Cells
  • Cell Size Analysis & Size-based Counting
  • Applications:
    • Adoptive Cell Transfer Therapy
    • Cell Viability Measurement Using Trypan Blue or AO/PI
    • Cell Size Assay
    • NCI-60 Cancer Cell Lines
    • Insect Cells
    • Stem Cells
Specifications
Camera Bright field imaging and pattern-recognition software
For Use With (Equipment) Cellometer Counting Chambers
Includes Cellometer Mini Instrument, Cellometer Software, Power supply, USB 2.0 Connection Cable, and Phone/online applications support during set-up
Depth (Metric) 19.8 cm
Height (Metric) 11.9 cm
Weight (Metric) 2.0 kg
Width (Metric) 10.9 cm