Learn More
Nexcelom™ Cellometer Disposable Counting Chambers (PD100 Slides)
Product Code. 11522186
Encompass
Change view
Click to view available options
Volume (Metric) Working: 20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11522186 Volume (Metric) Working: 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11522186 Supplier Nexcelom™ Supplier No. CHT4003
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
The fixed 20 μL sample size allows for simple, automated calculation of cell concentration following imaging and counting port
  • Each all-plastic, disposable slide contains two sample counting chambers with precisely controlled height
  • After loading the sample, the slide is inserted into a Cellometer System
  • Cells are imaged directly from the center of the counting chamber
  • Image-based counting with disposable counting chambers offers several key advantages:
    • No potential clogging
    • Ideal for fragile samples, such as hepatocytes
    • No washing
    • No potential cross-contamination
Specifications
Volume (Metric) Working 20 μL
Packaging 10 individual boxes
No. per Pack 500