missing translation for 'onlineSavingsMsg'
Learn More

Cellobiose, Millipore™

Product Code. 10683221
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10683221 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MQ100 Graded - For Non-regulated Laboratory Applications
Product Code. 10683221 Supplier Millipore Supplier No. 1023520025
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ100 Graded - For Non-regulated Laboratory Applications

For biochemistry

TRUSTED_SUSTAINABILITY

Specifications

CAS 528-50-7
Format Bottle
Melting Point 225°C to 230°C (Decomposition)
Quantity 25 g
Synonym Cellobiose, Cellobiose
Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.