Learn More
Invitrogen™ CellMask™ Actin Tracking Stain Variety Pack
Product Code. 17113279
Change view
Click to view available options
Quantity:
3 x 60 Assays
Unit Size:
Pack of 180
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17113279 3 x 60 Assays Pack of 180
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17113279 Supplier Invitrogen™ Supplier No. A57249
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Stain intracellular F-actin with the CellMask Actin Tracking Stain Variety Pack in live or fixed cells ICC, IHC, and 3D cell culture. The stains easily permeate cell membranes, are retained within cells after loading, and show no detectable effects on viability after 24 hours of incubation.
Stain and track intracellular filamentous actin (F-actin) with the CellMask Actin Tracking Stain Variety Pack in live or fixed cells in immunofluorescence (IF), immunocytochemistry (ICC), and immunohistochemistry (IHC) protocols, and in 3D cell culture. The CellMask Actin Tracking Stain Variety Pack contains one vial each of CellMask Green, Orange, and Deep Red Actin Tracking Stains, for fluorescent labeling of polymerized/F-actin in live or fixed cells. Because the stains are designed to readily permeate live cells, they produce bright and uniform labeling results. The CellMask Actin Tracking Stain Variety Pack can be used to track F-actin in live cells for 24 hours or more. Live cells stained with the kit's tracking stains can be fixed; alternately, the stains can be used on already fixed cells. The Cell Mask Actin Tracking Stain Variety Pack can be multiplexed with antibodies in IF, ICC, and IHC protocols, and can also be used in 3D cell culture for 3D imaging.
Features of Cell Mask actin tracking stains include:
  • Stain F-actin in live or fixed cells for fluorescence 2D monolayer or 3D imaging experiments
  • Can be multiplexed with antibodies in IF/ICC/IHC experiments
CellMask actin tracking stains are also available individually in three colors: green (Cat. No. A57243, A57246) detected with a FITC/GFP filter (Ex 503 nm/Em 512 nm), orange (Cat. No. A57244, A57247), detected with a TRITC/RFP filter (Ex 545 nm/Em 570 nm), and deep red (Cat. No. A57245, A57248) detected with a CY5/Deep Red filter (Ex 652 nm/Em 669 nm).
CellMask actin tracking stains detect F-actin with a targeting structure that closely resembles jasplakinolide. They do not label globular actin (G-actin) or monomer actin (M-actin). CellMask actin tracking stains easily permeate live cell membranes and are retained within live cells after loading. As demonstrated in the figures below, incubation of cells with CellMask actin tracking stains has no detectable effect on the viability of various cell lines after 24 hours of incubation.
CellMask actin tracking stains detect F-actin in live cells or tissue, which can then be fixed. In addition, CellMask actin tracking stains can be used to detect F-Actin in paraformaldehyde fixed cells or tissue (freshly fixed or cryo frozen only) and can be multiplexed with antibody detection.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage • CellMask Green Actin Tracking stain, 1 vial
• CellMask Orange Actin Tracking stain, 1 vial
• CellMask Deep Red Actin Tracking stain, 1 vial
Each vial contains enough reagent to stain 120 slides, when 500 μL of staining solution is used per slide
For Use With (Equipment) Fluorescence Microscope, High Content Analyzer
Quantity 3 x 60 Assays
Detection Method Fluorescence Microscopy
Description CellMask™ Actin Tracking Stain Variety Pack
Product Line CellMask
Shipping Condition Room Temperature
Target F-Actin
Label Type Fluorescent Dye
Product Type Actin
Sub Cellular Localization Actin, Cytoskeleton