Learn More
Thermo Scientific™ CellInsight™ CX7 High Content Analysis Platform
Shop All Thermo Scientific Products
Description
Integrated system for high content analysis
The Thermo Scientific™ CellInsight™ CX7 High Content Analysis (HCA) Platform is a fast, automated cellular imaging and analysis platform designed for quantitative microscopy.
The CellInsight CX7 HCA Platform offers an integrated way to develop and automate high content assays using a combination of 5-color brightfield and 7-color fluorescence imaging in confocal or wide-field modes. See how the CellInsight CX7 platform compares to other high content analysis platforms from Thermo Fisher Scientific.
Features of the CellInsight CX7 HCA Platform
Capabilities
The CellInsight CX7 HCA Platform offers an integrated way to develop and automate high content assays using a combination of 5-color brightfield and 7-color fluorescence imaging in confocal or wide-field modes. See how the CellInsight CX7 platform compares to other high content analysis platforms from Thermo Fisher Scientific.
Features of the CellInsight CX7 HCA Platform
- 7-channel fluorescent imaging
- 5-channel bright-field imaging
- 7-channel confocal imaging
- Software and laser-based autofocus for consistent scan times
- Configured for fully automated plate handling and scanning
Capabilities
- Addresses hundreds of assay types with flexible software and hardware tools
- Scan and analyze microscope slides or most SBS-standard format microplates
- Use HCS Studio software for integrated instrument control and data analysis
- Achieve the fastest time to results by using intelligent acQuisition (iQ) real-time analysis feedback
- Compatible with Molecular Probes™ fluorescent detection reagents
- Stable and reliable light source enables reduced variability, increased data quality, and decreased operating costs
- Broad reagent options with seven-color excitation from violet to near IR for assay multiplexing
- Dual mode focusing using Software and Laser Autofocus methods to address both labelled and unlabeled sample types
- Multi-mode imaging allows for wide-field, bright-field, and confocal imaging in one run
- HCS Studio Cell Analysis Software is an integrated instrument control, visualization, and analysis software package for novice to advanced users
- Scalable with Store Image and Database Software and optional Orbitor RS Microplate handler
Specifications
Specifications
| Product Line | CellInsight |
| Description | CellInsight™ CX7 High Content Analysis Platform |
| For Use With (Equipment) | High Content Instrument |
| Color | Assorted Colors |
| Detection Method | Chromogenic, Fluorescence |
| Format | Microplate, Slide(s) |
| Content And Storage | • 1 Reader |
| Shipping Condition | Room Temperature |