Learn More
Corning™ cellgro™ IMDM Liquid Classical Cell Culture Media
BIOPHARMA
Product Code. 15313601
Change view
Click to view available options
Quantity:
500 mL
1 L
Formulation:
Inorganic Salts: 165.00mg/L CaCl2 (anhydrous), 330.00mg/L KCl, 0.076mg/L KNO3, 97.70mg/L MgSO4 (anhydrous), 0.0173mg/L Na2SeO3, 4,505.00mg/L NaCl, 125.00mg/L NaH2PO4.H2O, 3,024.00mg/L NaHCO3; Amino Acids: 25.00mg/L L-Alanine, 84.00mg/L L-Arginine. HCl, 28.40mg/L L-Asparagine.H2O, 30.00mg/L L-Aspartic acid, 91.24mg/L L-Cystine.2HCl, 75.00mg/L L-Glutamic acid, 584.00mg/L L-glutamine, 30.00mg/L Glycine, 42.00mg/L L-Histidine.HCl.H2O, 105.00mg/L L-Isoleucine, 105.00mg/L L-Leucine, 146.00mg/L L-Lysine. HCl, 30.00mg/L L-Methionine, 66.00mg/L L-Phenylalanine, 40.00mg/L L-Proline, 42.00mg/L L-Serine, 95.00mg/L L-Threonine, 16.00mg/L L-Tryptophan, 103.79mg/L L-Tyrosine.2Na.2H2O, 94.00mg/L L-Valine; Vitamins: 0.013mg/L Biotin, 4.00mg/L D-Calcium pantothenate, 4.00mg/L Choline chloride, 4.00mg/L Folic acid, 7.20mg/L i-Inositol, 4.00mg/L Nicotinamide, 4.00mg/L Pyridoxine. HCl, 0.40mg/L Riboflavin, 4.00mg/L Thiamine. HCl, 0.013mg/L Vitamin B12; Other: 4,500.00mg/L D-Glucose, 5,958.00mg/L HEPES, 15.00mg/L Phenol red. Na, 110.00mg/L Sodium pyruvate.
Sodium Pyruvate, Phenol Red, Pyridoxine HCl, Sodium Bicarbonate
Unit Size:
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Formulation unitSize
15313601 1 L Inorganic Salts: 165.00mg/L CaCl2 (anhydrous), 330.00mg/L KCl, 0.076mg/L KNO3, 97.70mg/L MgSO4 (anhydrous), 0.0173mg/L Na2SeO3, 4,505.00mg/L NaCl, 125.00mg/L NaH2PO4.H2O, 3,024.00mg/L NaHCO3; Amino Acids: 25.00mg/L L-Alanine, 84.00mg/L L-Arginine. HCl, 28.40mg/L L-Asparagine.H2O, 30.00mg/L L-Aspartic acid, 91.24mg/L L-Cystine.2HCl, 75.00mg/L L-Glutamic acid, 584.00mg/L L-glutamine, 30.00mg/L Glycine, 42.00mg/L L-Histidine.HCl.H2O, 105.00mg/L L-Isoleucine, 105.00mg/L L-Leucine, 146.00mg/L L-Lysine. HCl, 30.00mg/L L-Methionine, 66.00mg/L L-Phenylalanine, 40.00mg/L L-Proline, 42.00mg/L L-Serine, 95.00mg/L L-Threonine, 16.00mg/L L-Tryptophan, 103.79mg/L L-Tyrosine.2Na.2H2O, 94.00mg/L L-Valine; Vitamins: 0.013mg/L Biotin, 4.00mg/L D-Calcium pantothenate, 4.00mg/L Choline chloride, 4.00mg/L Folic acid, 7.20mg/L i-Inositol, 4.00mg/L Nicotinamide, 4.00mg/L Pyridoxine. HCl, 0.40mg/L Riboflavin, 4.00mg/L Thiamine. HCl, 0.013mg/L Vitamin B12; Other: 4,500.00mg/L D-Glucose, 5,958.00mg/L HEPES, 15.00mg/L Phenol red. Na, 110.00mg/L Sodium pyruvate. Pack of 6
15343531 500 mL Inorganic Salts: 165.00mg/L CaCl2 (anhydrous), 330.00mg/L KCl, 0.076mg/L KNO3, 97.70mg/L MgSO4 (anhydrous), 0.0173mg/L Na2SeO3, 4,505.00mg/L NaCl, 125.00mg/L NaH2PO4.H2O, 3,024.00mg/L NaHCO3; Amino Acids: 25.00mg/L L-Alanine, 84.00mg/L L-Arginine. HCl, 28.40mg/L L-Asparagine.H2O, 30.00mg/L L-Aspartic acid, 91.24mg/L L-Cystine.2HCl, 75.00mg/L L-Glutamic acid, 584.00mg/L L-glutamine, 30.00mg/L Glycine, 42.00mg/L L-Histidine.HCl.H2O, 105.00mg/L L-Isoleucine, 105.00mg/L L-Leucine, 146.00mg/L L-Lysine. HCl, 30.00mg/L L-Methionine, 66.00mg/L L-Phenylalanine, 40.00mg/L L-Proline, 42.00mg/L L-Serine, 95.00mg/L L-Threonine, 16.00mg/L L-Tryptophan, 103.79mg/L L-Tyrosine.2Na.2H2O, 94.00mg/L L-Valine; Vitamins: 0.013mg/L Biotin, 4.00mg/L D-Calcium pantothenate, 4.00mg/L Choline chloride, 4.00mg/L Folic acid, 7.20mg/L i-Inositol, 4.00mg/L Nicotinamide, 4.00mg/L Pyridoxine. HCl, 0.40mg/L Riboflavin, 4.00mg/L Thiamine. HCl, 0.013mg/L Vitamin B12; Other: 4,500.00mg/L D-Glucose, 5,958.00mg/L HEPES, 15.00mg/L Phenol red. Na, 110.00mg/L Sodium pyruvate. Pack of 6
15373551 500 mL Sodium Pyruvate, Phenol Red, Pyridoxine HCl, Sodium Bicarbonate Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15313601 Supplier Corning™ Supplier No. 10016CM
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
To assess growth promotion and cytotoxicity characteristics, Corning Classical Media products undergo a variety of tests to determine functionality and toxicity using growth-promotion analysis. Product performance is measured by harvest-to-plant ratios or fold increase according to predetermined specifications. Liquid and powder classical media products are also tested to determine pH and osmolality. Classical media powders are also tested for residual moisture. The pH and osmolality tests are performed according to specific procedures in which all equipment is calibrated using standards traceable to the National Institute of Standards and Technology. Biological testing of Corning liquid classical media products guarantees low levels of endotoxin, mycoplasma, and an absence of contamination. Endotoxins: classical media products are tested for bacterial endotoxin using the Limulus Amoebocyte Lysate (LAL) Chromogenic Assay. For release, each lot of standard liquid classical media must have an endotoxin level of less than 0.25 EU/mL. Classical media powder products must have an endotoxin level of less than 0.25 EU/mL when reconstituted to formula weight with low endotoxin water. All Corning liquid classical media products are tested for sterility and liquid classical media products are tested for mycoplasma off-site using a large-volume method.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 1 X
Product Type IMDM
For Use With (Application) To assess growth promotion and cytotoxicity characteristics
Quantity 1 L
Endotoxin Level <0.25EU/mL
Formulation Inorganic Salts: 165.00mg/L CaCl2 (anhydrous), 330.00mg/L KCl, 0.076mg/L KNO3, 97.70mg/L MgSO4 (anhydrous), 0.0173mg/L Na2SeO3, 4,505.00mg/L NaCl, 125.00mg/L NaH2PO4.H2O, 3,024.00mg/L NaHCO3; Amino Acids: 25.00mg/L L-Alanine, 84.00mg/L L-Arginine. HCl, 28.40mg/L L-Asparagine.H2O, 30.00mg/L L-Aspartic acid, 91.24mg/L L-Cystine.2HCl, 75.00mg/L L-Glutamic acid, 584.00mg/L L-glutamine, 30.00mg/L Glycine, 42.00mg/L L-Histidine.HCl.H2O, 105.00mg/L L-Isoleucine, 105.00mg/L L-Leucine, 146.00mg/L L-Lysine. HCl, 30.00mg/L L-Methionine, 66.00mg/L L-Phenylalanine, 40.00mg/L L-Proline, 42.00mg/L L-Serine, 95.00mg/L L-Threonine, 16.00mg/L L-Tryptophan, 103.79mg/L L-Tyrosine.2Na.2H2O, 94.00mg/L L-Valine; Vitamins: 0.013mg/L Biotin, 4.00mg/L D-Calcium pantothenate, 4.00mg/L Choline chloride, 4.00mg/L Folic acid, 7.20mg/L i-Inositol, 4.00mg/L Nicotinamide, 4.00mg/L Pyridoxine. HCl, 0.40mg/L Riboflavin, 4.00mg/L Thiamine. HCl, 0.013mg/L Vitamin B12; Other: 4,500.00mg/L D-Glucose, 5,958.00mg/L HEPES, 15.00mg/L Phenol red. Na, 110.00mg/L Sodium pyruvate.
Content And Storage 2 to 8°C