Learn More
Invitrogen™ Celleste™ 2D Deconvolution Module, single-user license
Product Code. 16606484
Change view
Click to view available options
Quantity:
1 card
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16606484 1 card Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16606484 Supplier Invitrogen™ Supplier No. AMEP4864
 Restricted Item
Estimated Shipment: 05-11-2026
Suite of image analysis and visualization tools designed for a range of biological applications that lets you transform your images into quantitative data in a streamlined and customizable workflow
  • Celleste 5.0 Image Analysis Software is a suite of image analysis and visualization tools designed for a range of biological applications that lets you transform your images into quantitative data in a streamlined and customizable workflow. Coupled with the powerful image acquisition capabilities of our EVOS microscopes, Celleste 5 software allows you to seamlessly capture, process, measure, analyze, and share images and data.
  • Optional modules for 2D and 3D deconvolution, visualization, and analysis allow you to customize the capabilities according to your needs. This Celleste 5 2D Blind/Non-Blind Deconvolution Module helps improve single-plane image quality (S/N) of cells or tissue slices by clearing background haze (out-of-focus light)
  • Streamlined workflow tools that make it easy to visualize and edit images
  • Powerful image analysis capabilities for segmentation, classification, and quantification of single images or a batch of images
  • Blind and non-blind deconvolution options
  • Dramatically improved image quality
  • Removal of blur that can obscure important details
  • The tools to generate more impressive images and reliable data
  • Deconvolution is an algorithm-based process used to restore systematic loss of image contrast (blur) that results from the image point spread function (PSF). The powerful 2D and 3D restoration algorithms available in the Celleste 5 modules allow you to deconvolve your images to enhance contrast and quality, or to visualize them in Z, over time and by channel, then analyze the resulting image using an expanded tool set.
TRUSTED_SUSTAINABILITY
Specifications
Product Line Invitrogen
For Use With (Equipment) EVOS
Quantity 1 card
Shipping Condition Room Temperature