missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One CELLCOAT™ Collagen Type I Coated Cell Culture Flask

Product Code. 11709492
Change view
Click to view available options
Culture Area:
175 cm2
75 cm2
Unit Size:
Pack of 40
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Culture Area unitSize
11709492 175 cm2 Pack of 40
13092263 75 cm2 Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
An item in this group is an exact match for part number 07000196
Product Code. 11709492 Supplier Greiner Bio-One Supplier No. 661950
Add to basket
Add to basket
This item is not returnable. View return policy
An item in this group is an exact match for part number 07000196

Ideal for cell cultivation in serum-free or serum-reduced medium

  • Promotion of cell adhesion, proliferation and growth of endothelial cell, hepatocytes, muscle cells, pheochromocytoma cells (PC12) and other cell types
  • Source: Rat tail Collagen Type I
  • Ready-to-use
  • Supplied with red filter screw cap.
TRUSTED_SUSTAINABILITY

Specifications

Closure Color Red
Closure Material HDPE
Capacity (Metric) 650 mL
Material Polystyrene
Neck Style Canted
No. per Case 40
No. per Pack 5
Surface Coating Collagen Type I Protein
Graduated Yes
Closure Type Filter Screw Cap
Culture Area 175 cm2
Volume (Metric) Working 20 to 85 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.