Learn More
Greiner Bio-One CELLCOAT™ Collagen Type 1-Coated Multiwell Plates
Product Code. 10207401
Change view
Click to view available options
Well Count:
6
24
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Well Count unitSize
10207401 24 Pack of 50
11789512 6 Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10207401 Supplier Greiner Bio-One Supplier No. 662950
Add to basket
Add to basket
Suitable for serum-free and serum-reduced cell cultivation and transfection
  • Coated with Collagen 1
  • Improved adhesion and proliferation of primary cells, various cell lines
  • Sterile
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Autoclavable Not Autoclavable
Bottom Shape Flat
Capacity (Metric) 3.3 mL
Color Clear
Culture Area 1.9 sq. cm
Material Polystyrene
Shape Rectangular
Sterility Non-sterile
Surface Treatment Collagen Type I Protein
Type Multi-well Cell Culture Plate
Volume (Metric) Working 0.5 to 1.0 mL
Well Count 24
Includes Lid
No. per Case 50
No. per Pack 5
Temperature Range (Metric) Room Temperature
Dimensions (L x W x H) 128 x 85 x 19 mm
Length (Metric) 127.76 mm
Width (Metric) 85.48 mm
Height (Metric) 19 mm