Learn More
Cell Viability Incorporated™ CCD Cell Viability Reagent Pack
Product Code. 12740616
Change view
Click to view available options
Product Type:
Cell Viability Reagent Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
12740616 Cell Viability Reagent Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12740616 Supplier Cell Viability Incorporated™ Supplier No. 2400102
  • ONLINE EXCLUSIVE PRICE
    283.00 EUR
    153.04 EUR / Pack of 1
    Save 129.96 EUR
    45% Off
 Restricted Item
Estimated Shipment: 08-01-2027
Save money without losing performance with alternative reagents for automated CCD cell viability analyzers. Cell Viability Incorporated™ CCD Cell Viability Reagent Pack features long-term material stability.
  • Same count performance as original reagent pack vendor.
  • Suitable for Vi-Cell™ analyzers.
  • Long-term material stability - no cooling necessary.
  • Cost savings.
Specifications
For Use With (Equipment) Designed as a direct replacement for the Vi-Cell™ Analyzer reagent pack.
Description Includes reagent kit and 200 sample cups.
Product Type Cell Viability Reagent Kit
compliance-icons
Product Identifier
  • Cell Viability Reagent Pack
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
Supplemental information
  • MIXTURE LIST-Contain: isopropyl alcohol