Learn More
TPP™ Rotatable Cell Scrapers
Product Code. 11371714
Change view
Click to view available options
Length (Metric) Handle:
240 mm
Unit Size:
Pack of 150
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Handle unitSize
11371714 240 mm Pack of 150
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11371714 Supplier TPP™ Supplier No. 99002
Spotlight Offers
Use promo code 28036
View all promotions
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
Designed with sharp edge blade to collect clean and complete cell sample. TPP™ Rotatable Cell Scrapers feature a free rotating blade to provide access to all corners.
To change blade angle: apply slight pressure pushing the handle downward towards the floor of the container; slight rotation of the handle will cause the blade to twist to the desired direction.
  • Small raised knobs on the handle for a slip free and secure grip with rubber gloves
  • Lint-free blister packaging
  • Single wrapped, lint-free paper/plastick blister packaging
  • Convenient, stackable dispenser box 
Specifications
Width (Metric) Blade 13 mm
For Use With (Equipment) Tissue culture flasks, flat tubes, dishes
Length (Metric) Handle 240 mm
Material Polyethylene