missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hellma™ Cell Holders
Click to view available options
For Use With (Equipment):
Cell type 106
Support For 4 Spectro Tanks
Unit Size:
Each
Specifications
Specifications
| Dimensions (L x W x H) | 45 x 12.5 x 12.5 mm |
| For Use With (Equipment) | Cell type 106 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction