Learn More
Cole-Parmer™ Essentials Replacement Cell Culturing Flask
Product Code. 12937286
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12937286 250 mL Each
12927286 125 mL Each
11709302 500 mL Each
12947286 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12937286 Supplier Cole-Parmer™ Essentials Supplier No. 2183735
  • ONLINE EXCLUSIVE PRICE
    339.00 EUR
    311.81 EUR / Pack of 1
    Save 27.19 EUR
    8% Off
In Stock
Add to basket
Edge
Add to basket
Enhance the life of your BS-200 series cell culture stirring systems with this Replacement Cell Culturing Flask.
  • Constructed of borosilicate glass
The culture vessels incorporate a unique base design which, together with the bulb-ended stirrer, ensures the cells are lifted into suspension at the lowest possible speeds. This gentle stirring action promotes high cell yields by preventing cell shearing. Replacement flasks do not include caps or stirring mechanisms.
Specifications
Capacity (Metric) 250 mL
Material Borosilicate Glass
For Use With (Equipment) Biological Stirrers Glass no End Caps, Seals or Stirrer Rods
Bottom Shape Flat