Learn More
Cole-Parmer™ Cell counting slide
Product Code. 11709679 Shop All Cole Parmer Products
Change view
Click to view available options
Product Type:
Cell Counting Slide
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11709679 Cell Counting Slide Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11709679 Supplier Cole-Parmer™ Supplier No. 0549160
  • ONLINE EXCLUSIVE PRICE
    102.00 EUR
    88.12 EUR / Pack of 1
    Save 13.88 EUR
    13% Off
View all promotions
Estimated Shipment: 09-02-2027
Specifications
Product Type Cell Counting Slide
Capacity (Metric) 1 mL
Quantity 1 Ea.
Height (English) 0.787 in.
Width (English) 1.96 in.