Learn More
Celestolide, TRC
Product Code. 30137975
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137975 100 mg 100mg
30132781 250 mg 250mg
17964375 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137975 Supplier Toronto Research Chemicals Supplier No. TRCC253500100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13171-00-1
Molecular Formula C17 H24 O
Molecular Weight (g/mol) 244.37
Synonym Ethanone, 1-[6-(1,1-dimethylethyl)-2,3-dihydro-1,1-dimethyl-1H-inden-4-yl]- (9CI, ACI), 1-[6-(1,1-Dimethylethyl)-2,3-dihydro-1,1-dimethyl-1H-inden-4-yl]ethanone (ACI), Ketone, 6-tert-butyl-1,1-dimethyl-4-indanyl methyl (6CI, 7CI, 8CI), 1-(6-(tert-Butyl)-1,1-dimethyl-2,3-dihydro-1H-inden-4-yl)ethanone, 1-(6-tert-Butyl-1,1-dimethyl-indan-4-yl)-ethanone, 1-[6-(tert-Butyl)-1,1-dimethyl-2,3-dihydro-1H-inden-4-yl]ethan-1-one, 4-Acetyl-1,1-dimethyl-6-tert-butylindan, 4-Acetyl-1,1-dimethyl-6-tert-butylindane, 4-Acetyl-6-tert-butyl-1,1-dimethylindan, 4-Acetyl-6-tert-butyl-1,1-dimethylindane, 6-tert-Butyl-1,1-dimethyl-4-indanyl methyl ketone, ADBI, Celestolide, Crysolide, Esperone
IUPAC Name 1-(6-tert-butyl-1,1-dimethyl-2,3-dihydroinden-4-yl)ethanone
SMILES CC(=O)c1cc(cc2c1CCC2(C)C)C(C)(C)C
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C17H24O/c1-11(18)14-9-12(16(2,3)4)10-15-13(14)7-8-17(15,5)6/h9-10H,7-8H2,1-6H3
Formula Weight 244.1827
Chemical Name or Material Celestolide