missing translation for 'onlineSavingsMsg'
Learn More

Cefteram, TRC

Product Code. 17974375
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17974375 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17974375 Supplier Toronto Research Chemicals Supplier No. TRCC2442755MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82547-58-8
Molecular Formula C16 H17 N9 O5 S2
Molecular Weight (g/mol) 479.49
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2Z)-2-(2-amino-4-thiazolyl)-2-(methoxyimino)acetyl]amino]-3-[(5-methyl-2H-tetrazol-2-yl)methyl]-8-oxo-, (6R,7R)-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2-amino-4-thiazolyl)(methoxyimino)acetyl]amino]-3-[(5-methyl-2H-tetrazol-2-yl)methyl]-8-oxo-, [6R-[6α,7β(Z)]]-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2Z)-(2-amino-4-thiazolyl)(methoxyimino)acetyl]amino]-3-[(5-methyl-2H-tetrazol-2-yl)methyl]-8-oxo-, (6R,7R)- (9CI), (6R,7R)-7-[[(2Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetyl]amino]-3-[(5-methyl-2H-tetrazol-2-yl)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, Antibiotic Ro 19-5247, Antibiotic T 2525, Cefteram, Cefterame, Ceftetrame, Ro 19-5247, T 2525;
IUPAC Name (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-[(5-methyltetrazol-2-yl)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)O)Cn3nnc(C)n3)\c4csc(N)n4

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C16H17N9O5S2/c1-6-20-23-24(21-6)3-7-4-31-14-10(13(27)25(14)11(7)15(28)29)19-12(26)9(22-30-2)8-5-32-16(17)18-8/h5,10,14H,3-4H2,1-2H3,(H2,17,18)(H,19,26)(H,28,29)/b22-9-/t10-,14-/m1/s1
Formula Weight 479.0794
Chemical Name or Material Cefteram
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.