Learn More
Cediranib, MedChemExpress
Product Code. 16473698
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
Product Code. 16473698
Brand: Medchem Express HY10205
MedChemExpress Cediranib (AZD2171) is a highly potent, orally available VEGFR tyrosine kinase inhibitor with IC50s of <1, <3, 5, 5, 36, 2 nM for Flt1, KDR, Flt4, PDGFRα, PDGFRβ, c-Kit, respectively.
Cediranib (AZD2171) is a highly potent, orally available VEGFR tyrosine kinase inhibitor with IC50s of <1, <3, 5, 5, 36, 2 nM for Flt1, KDR, Flt4, PDGFRα, PDGFRβ, c-Kit, respectively.
Specifications
Chemical Name or Material Cediranib
CAS 288383-20-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H27FN4O3
Quantity 10 mg
Synonym AZD2171
Solubility Information DMSO : 20 mg/mL (44.39 mM; Need ultrasonic)
SMILES FC1=C(OC2=C(C(C=C3OCCCN4CCCC4)=NC=N2)C=C3OC)C=CC5=C1C=C(C)N5
Molecular Weight (g/mol) 450.51
Formula Weight 450.51
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease