Learn More
Vileda Professional™ CE MicroIntensive Duo™
Product Code. 17812476
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Unit Size
17812476 Non-sterile Pack of 25
17822476 Sterile Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17812476 Supplier Vileda Professional™ Supplier No. 166813
  • ONLINE EXCLUSIVE PRICE
    400.00 EUR
    348.80 EUR / Pack of 25
    Save 51.20 EUR
    12% Off
Estimated Shipment: 18-02-2027
Double-sided, single-use mop to speed-up the cleaning process, while also reducing the risk of cross-contamination.
  • Independent lab study, conducted by BMA Labs, resulting in bacteria reduction of 99.99%
  • 4-layer construction - featuring Polyester/Microfiber embossed cleaning side
  • Pre-laundered for single-use
  • Gamma and autoclave compatible
  • Extremely low particle shedding
  • Double-sided cleaning with color -stitched side indicator
  • Corner drainage openings for cleaner rinses
  • Finger loop for touchless on/off of cleaning sides
Specifications
Product Type Cleanroom Mop
Color White
Sterility Non-sterile
Autoclavable Autoclavable
Material Microfiber Polyester