missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CDT2 Polyclonal antibody specifically detects CDT2 in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | CDT2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | CDT2RA-regulated nuclear matrix-associated protein, CDW1, DCAF2DDB1 and CUL4 associated factor 2, DDB1- and CUL4-associated factor 2, denticleless homolog (Drosophila), L2DTLRA regulated nuclear matrix associated protein, Lethal(2) denticleless protein homolog, RAMPdenticleless protein homolog, Retinoic acid-regulated nuclear matrix-associated protein |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 541-730 of human CDT2 (NP_057532.3).,, Sequence:, AEACSESRNRVKRRLDSSCLESVKQKCVKSCNCVTELDGQVENLHLDLCCLAGNQEDLSKDSLGPTKSSKIEGAGTSISEPPSPISPYASESCGTLPLPLRPCGEGSEMVGKENSSPENKNWLLAMAAKRKAENPSPRSPSSQTPNSRRQSGKTLPSPVTITPSSMRKICTYFHRKSQEDFCGPEHSTEL |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?