missing translation for 'onlineSavingsMsg'
Learn More

CDK4/6-IN-2, MedChemExpress

Product Code. 16461077
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16461077 50 mg 50mg
16424818 10 mg 10mg
16407567 5 mg 5mg
16405616 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16461077 Supplier Medchem Express Supplier No. HY114339

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CDK4/6-IN-2 is a potent CDK4 and CDK6 inhibitor extracted from patent US20180000819A1, Compound 1, has IC50s of 2.7 and 16 nM for CDK4 and CDK6, respectively.

CDK4/6-IN-2 is a potent CDK4 and CDK6 inhibitor extracted from patent US20180000819A1, Compound 1, has IC50s of 2.7 and 16 nM for CDK4 and CDK6, respectively.

Specifications

Chemical Name or Material CDK4/6-IN-2
CAS 1800506-48-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H32F2N8
Quantity 50 mg
Solubility Information DMSO : 5.2 mg/mL (10.26 mM; Need ultrasonic)
SMILES CCN1CCN(CC2=CN=C(NC3=NC=C(F)C(C4=CC(N(C(C)C)C(C)=N5)=C5C(F)=C4)=C3)N=C2)CC1
Molecular Weight (g/mol) 506.59
Formula Weight 506.59
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.