missing translation for 'onlineSavingsMsg'
Learn More

CD83 Antibody (MM0179-4B34) - Azide and BSA Free, Novus Biologicals™

Product Code. 18046297 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18046297 0.025 mg 25µg
18019636 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18046297 Supplier Novus Biologicals Supplier No. NBP2121800.025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD83 Monoclonal specifically detects CD83 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD83
Applications Flow Cytometry, CyTOF, Immunohistochemistry (Frozen), Immunohistochemistry
Classification Monoclonal
Clone MM0179-4B34
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:20-100, CyTOF-ready
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. Q01151
Gene Alias B-cell activation protein, BL11CD83 antigen, CD83 antigen (activated B lymphocytes, immunoglobulin superfamily), CD83 molecule, Cell surface protein HB15, cell-surface glycoprotein, HB15hCD83
Gene Symbols CD83
Host Species Mouse
Immunogen Human CD83
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Dendritic Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9308
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.