Learn More
IBA Lifesciences CD81 Fab-TACS™ Exosome Agarose Column Starter Kit
Shop All IBA Lifesciences ProductsDescription
- Fab-TACS™ is a traceless affinity exosome isolation approach based on our Strep-tag™ technology. CD81 Fab-TACS™ Exosome Agarose Column Starter Kit allows label-free isolation of human CD81+ exosomes from plasma, serum and cell culture supernatants. CD81 is a member of the tetraspanin family present on the surface of exosomes
- Strep-Tactin™ TACS Agarose columns are filled with Strep-Tactin™ coated agarose macrobeads developed for cell isolation. Fab fragments fused to a Twin-Strep-tag™ (Fab-Streps) bind to the matrix
- Exosomes adhere to the matrix based on the exclusive binding of the Fab-Strep to a surface marker. Non-target particles are efficiently washed away
- In a final step, the addition of biotin causes the elution of the exosomes and the Fab-Streps. After elution, the Fab-Streps self-dissociate from the exosome surface due to their low affinity
- The label-free, fully functional exosomes are now ready for further downstream applications
Specifications
Specifications
| Includes | Strep-Tactin™ TACS Agarose columns, 0.3 mL (2 x), CD81 Fab-Strep (human), 10 x Buffer CI, Biotin stock solution, TACS Column Adapter |
| Target Species | Human |
For Research Use Only.
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?