missing translation for 'onlineSavingsMsg'
Learn More

CD69 Antibody (H1.2F3), PE, Novus Biologicals™

Product Code. 18272030 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.25 mL
Unit Size:
0.25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18272030 0.25 mL 0.25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18272030 Supplier Novus Biologicals Supplier No. NBP128011PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Armenian Hamster Monoclonal Antibody

CD69 Monoclonal specifically detects CD69 in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD69
Applications Flow Cytometry
Classification Monoclonal
Clone H1.2F3
Conjugate PE
Dilution Flow Cytometry
Gene Alias Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leukocyte surface antigen Leu-23, MLR-3
Gene Symbols CD69
Host Species Armenian Hamster
Immunogen Mouse dendritic epidermal cell line Y245
Purification Method Protein A or G purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 969
Test Specificity Mouse CD69/Very Early Activation Antigen (VEA), Mr 85 kDa (unreduced)
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.