missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse anti-Human , Alexa Fluor 647, Clone: OKT3, BD Biosciences™
Description
CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | CD3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OKT3 |
| Conjugate | Alexa Fluor 647 |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | CD3E; CD3e; T-cell surface antigen T3/Leu-4 epsilon; T3E; TCRE |
| Host Species | Mouse |
| Immunogen | Sheep Erythrocyte Rosette-purified Human T Cells |
| Quantity | 100 Tests |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?