missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD27 Ligand/TNFSF7/CD70 Antibody (TAN1-6) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
256.00 EUR - 705.00 EUR
Specifications
| Antigen | CD27 Ligand/TNFSF7/CD70 |
|---|---|
| Clone | TAN1-6 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18627156
|
Novus Biologicals
NBP3-09030-0.025mg |
0.025 mg |
256.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18665486
|
Novus Biologicals
NBP3-09030-0.2mg |
0.2 mg |
705.00 EUR
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD27 Ligand/TNFSF7/CD70 Monoclonal antibody specifically detects CD27 Ligand/TNFSF7/CD70 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, NeutralizationSpecifications
| CD27 Ligand/TNFSF7/CD70 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CD27-L, CD27LCD27 ligand, CD27LG, CD70, CD70 molecule, Ki-24 antigen, surface antigen CD70, TNFSF7CD70 antigen, tumor necrosis factor (ligand) superfamily, member 7, Tumor necrosis factor ligand superfamily member 7 | |
| This antibody was raised by immunising LOU rats with soluble recombinant CD70:Fc fusion protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| TAN1-6 | |
| Western Blot, Flow Cytometry, Immunofluorescence, Neutralization | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| PBS | |
| 970 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title