Learn More
CD27 Ligand/TNFSF7/CD70 Antibody (TAN1-6) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-09030-0.025mg
 View more versions of this product
Product Code. 18627156
  • 256.00 EUR / 25µg
Description
CD27 Ligand/TNFSF7/CD70 Monoclonal antibody specifically detects CD27 Ligand/TNFSF7/CD70 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Neutralization
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD27 Ligand/TNFSF7/CD70
Monoclonal
1 mg/mL
PBS
Rabbit
Protein A purified
RUO
Primary
Mouse
Purified
Western Blot, Flow Cytometry, Immunofluorescence, Neutralization
TAN1-6
Unconjugated
CD27-L, CD27LCD27 ligand, CD27LG, CD70, CD70 molecule, Ki-24 antigen, surface antigen CD70, TNFSF7CD70 antigen, tumor necrosis factor (ligand) superfamily, member 7, Tumor necrosis factor ligand superfamily member 7
This antibody was raised by immunising LOU rats with soluble recombinant CD70:Fc fusion protein
0.025 mg
Apoptosis
970
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions