missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD268 (BAFF Receptor) Monoclonal Antibody (8A7), Super Bright™ 436, eBioscience™, Invitrogen™

Product Code. 16391314
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
Unit Size:
Pack of 100
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16391314 25 Tests Pack of 25
16301324 100 Tests Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16391314 Supplier Invitrogen™ Supplier No. 62911741

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Tumor necrosis factor receptor superfamily member 13C (TNFRSF13C), also known as BAFFR, is a protein in humans is encoded by the TNFRSF13C gene. The BAFFR gene is mapped to chromosome 22q13. 1-q13. 31. It has got 184 amino acid transmembrane protein which is 56% identical to the mouse protein. B cell-activating factor (BAFF) enhances B-cell survival in vitro and is a regulator of the peripheral B-cell population. BAFF plays a crucial role in B cell development and can function through receptors other than BCMA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD268 (BAFF Receptor)
Applications Flow Cytometry
Classification Monoclonal
Clone 8A7
Concentration 5 μL/Test
Conjugate Super Bright 436
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene TNFRSF13C
Gene Accession No. Q96RJ3
Gene Alias 2010006P15Rik; B cell-activating factor receptor; BAFF receptor; BAFFR; BAFF-R; B-cell maturation defect; B-cell maturation defect 1; B-cell-activating factor receptor; Bcmd; Bcmd1; Bcmd-1; BLyS receptor 3; BR3; BROMIX; CD268; CVID4; Lvis22; prolixin; RGD1560810; TNF receptor superfamily member 13C; Tnfrsf13c; Tumor necrosis factor receptor superfamily member 13C; tumor necrosis factor receptor superfamily, member 13C
Gene Symbols TNFRSF13C
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115650
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.