missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD23, Rabbit Monoclonal Antibody

Product Code. 12623777 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
Unit Size:
100µL
12mL
1mL
500µL
7mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Form Unit Size
12623777 7 mL Prediluted 7mL
12673777 100 μL Concentrated 100µL
12693777 500 μL Concentrated 500µL
12653777 1 mL Concentrated 1mL
12643777 12 mL Ready-to-Use 12mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12623777 Supplier Epredia™ Supplier No. RM9123R7
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Obtain accurate and reproducible results in immunohistochemistry procedures with Epredia™ CD23, Rabbit Monoclonal Antibody.

CD23 is a 45 kDa glycoprotein, which is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. The CD23 molecule is identical to the low affinity IgE receptor found on B cells. Expression of CD23 has been detected in neoplastic cells from cases of B cell chronic lymphocyctic leukaemia and some cases of centroblastic/centrocytic lymphoma.

Host Species: Rabbit

Clone: SP23

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not known

Immunogen: Recombinant protein encoding human CD23 48-248aa

Molecular Weight: 45 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD23
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP23
Conjugate Unconjugated
Host Species Rabbit
Immunogen Recombinant protein encoding human CD23 48-248aa
Quantity 7 mL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.