missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCT8 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 EUR - 410.00 EUR
Specifications
| Antigen | CCT8 |
|---|---|
| Concentration | 0.15 mg/mL |
| Applications | Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16859873
|
Proteintech
12263-1-AP-20UL |
20 μL |
145.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16849873
|
Proteintech
12263-1-AP-150UL |
150 μL |
410.00 EUR
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MA1-82683 recognizes the theta polypeptide of the CCT chaperonin molecule complex. The intact CCT complex is composed of eight polypeptides in a double ring structure. CCT is important within cells in aiding the folding of proteins including actin, tubulin and the VHL tumor suppressor protein.Specifications
| CCT8 | |
| Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Mouse, Zebrafish, Rat, Human | |
| P42932, P50990 | |
| 10694, 12469, 288305, 394037 | |
| CCT8 Fusion Protein Ag2926 | |
| Primary | |
| -20°C | |
| CCT8 |
| 0.15 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| C21orf112, CCT theta, CCT8, Cctq, D21S246, KIAA0002, PRED71, TCP 1 theta | |
| Cct8 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title