missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCT8 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12263-1-AP-150UL
This item is not returnable.
View return policy
Description
MA1-82683 recognizes the theta polypeptide of the CCT chaperonin molecule complex. The intact CCT complex is composed of eight polypeptides in a double ring structure. CCT is important within cells in aiding the folding of proteins including actin, tubulin and the VHL tumor suppressor protein.Specifications
| CCT8 | |
| Polyclonal | |
| Unconjugated | |
| CCT8 | |
| C21orf112, CCT theta, CCT8, Cctq, D21S246, KIAA0002, PRED71, TCP 1 theta | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10694, 12469, 288305, 394037 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P42932, P50990 | |
| Cct8 | |
| CCT8 Fusion Protein Ag2926 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat, Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction