missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCT128930, MedChemExpress
Description
CCT128930 is a ATP-competitive and selective inhibitor of AKT (IC50=6 nM for AKT2). CCT128930 has 28-fold selectivity over the closely related PKA kinase (IC50=168 nM) through the targeting of Met282 of AKT (Met173 of PKA-AKT chimera), as well as 20-fold selectivity over p70S6K (IC50=120 nM). Antitumor activity.
Specifications
Specifications
| Chemical Name or Material | CCT128930 |
| CAS | 885499-61-6 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C18H20ClN5 |
| Quantity | 100 mg |
| Solubility Information | DMSO : 33.33 mg/mL (97.50 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | ClC1=CC=C(C=C1)CC2(CCN(CC2)C3=NC=NC4=C3C=CN4)N |
| Molecular Weight (g/mol) | 341.84 |
| Formula Weight | 341.84 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?