missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ccdc89 Polyclonal Antibody

Description
Immunogen sequence: PEERLEKQNEK LNNQEEETEF KELDGLREAL ANLRGLSEEE RSEKAMLRSR IEEQSQLICI LKRRSDEALE RCQILELLNA ELEEKMMQE Highest antigen sequence identity to the following orthologs - mouse 83%, rat 81%.
CCDC89 is a protein coding gene.
Specifications
Specifications
| Antigen | Ccdc89 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CCDC89 |
| Gene Accession No. | Q8N998 |
| Gene Alias | 1700019B01Rik; AU022100; bc8 orange-interacting protein; bc8 orange-interacting protein {ECO:0000303; BOIP; ccdc89; ccdc89 {ECO:0000312; coiled-coil domain containing 89; coiled-coil domain-containing protein 89; coiled-coil domain-containing protein 89 {ECO:0000312; ECO:0000312; EMBL:AAI67063.1, ECO:0000312; EMBL:AAI670631; PubMed:14648848}; RGD:1305335}; RGD1305335; zgc:172182 |
| Gene Symbols | CCDC89 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?