missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cbx8 Antibody (9C3D4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
203.00 EUR - 552.00 EUR
Specifications
| Antigen | Cbx8 |
|---|---|
| Clone | 9C3D4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18189738
|
Novus Biologicals
NBP2-37411 |
0.1 mL |
552.00 EUR
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18699035
|
Novus Biologicals
NBP2-37411-0.025ml |
0.025 mL |
203.00 EUR
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cbx8 Monoclonal specifically detects Cbx8 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| Cbx8 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| Unconjugated | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 57332 | |
| Purified recombinant fragment of human CBX8 (AA: 17-222) expressed in E. Coli. | |
| Primary | |
| 43.4 kDa |
| 9C3D4 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human | |
| chromobox homolog 8, chromobox homolog 8 (Drosophila Pc class), chromobox homolog 8 (Pc class homolog, Drosophila), hPc3, Pc3, PC3chromobox protein homolog 8, polycomb 3, Polycomb 3 homolog, RC1Pc class 3 homolog, Rectachrome 1 | |
| CBX8 | |
| IgG1 | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title