missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cbx8 Antibody (9C3D4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37411
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Cbx8 Monoclonal specifically detects Cbx8 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Tekniske data
| Cbx8 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| chromobox homolog 8, chromobox homolog 8 (Drosophila Pc class), chromobox homolog 8 (Pc class homolog, Drosophila), hPc3, Pc3, PC3chromobox protein homolog 8, polycomb 3, Polycomb 3 homolog, RC1Pc class 3 homolog, Rectachrome 1 | |
| Mouse | |
| 43.4 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| 9C3D4 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CBX8 | |
| Purified recombinant fragment of human CBX8 (AA: 17-222) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| 57332 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion