missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CBP20 Antibody, Novus Biologicals™

Product Code. 30047537 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30047537 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30047537 Supplier Novus Biologicals™ Supplier No. NBP329223

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CBP20 Polyclonal antibody specifically detects CBP20 in Human,Mouse samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBP20
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Tris-buffered Saline 0.1% BSA
Gene Alias 20 kDa nuclear cap-binding protein, Cbc2, CBP20CBC2, NCBP 20 kDa subunit, NCBP interacting protein 1, NCBP-interacting protein 1, NIP1Cell proliferation-inducing gene 55 protein, nuclear cap binding protein subunit 2, 20kD, nuclear cap binding protein subunit 2, 20kDa, nuclear cap-binding protein subunit 2
Host Species Rabbit
Immunogen Between 106 and 156
Purification Method Affinity purified
Quantity 10 μg
Regulatory Status RUO
Research Discipline Apoptosis, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 22916
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.