Learn More
Applied Biosystems™ Cathode Buffer Container Dx
Product Code. 15968826
Change view
Click to view available options
Quantity:
4 Ea.
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15968826 4 Ea. Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15968826 Supplier Applied Biosystems™ Supplier No. 4408258
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
The Cathode Buffer Container Dx contains 1X running buffer to support electrophoresis applications on Applied Biosystems Dx genetic analyzers.
The Cathode Buffer Container Dx is for use with Applied Biosystems Dx genetic analyzers.
The Cathode Buffer Container Dx contains 1X running buffer to support electrophoresis applications on Applied Biosystems Dx genetic analyzers. The Cathode Buffer Container Dx has two separate compartments: the left side provides the cathode buffer for electrophoresis and the right side provides for a capillary wash and spent polymer waste ejection functionality between injections. The Cathode Buffer Container Dx is a ready-to-use, disposable container with a radio frequency identification (RFID) tag incorporated into the label. The top of the Cathode Buffer Container Dx is heat sealed with a plastic film, which should be removed prior to direct installation on to the instrument. Each package includes four individual containers. The Septa Cathode Buffer Container Dx (sold separately) is used for sealing the Cathode Buffer Container Dx container.
This product is intended for in vitro diagnostic use on Applied Biosystems Dx genetic analyzers and is not validated for use with other systems or applications. Applied Biosystems Dx genetic analyzers are to be used by trained operators in the analysis of human DNA for the detection of fluorescently labeled DNA nucleotides by capillary electrophoresis. Refer to the appropriate Applied Biosystems Dx genetic analyzer user guide for further details.
The cathode buffer container (CBC) contains 1x running buffer to support all electrophoresis applications on the 3500 Dx series Genetic Analyzers.
Order Info
4 Pack
TRUSTED_SUSTAINABILITY
Specifications
Starting Material DNA, RNA
Quantity 4 Ea.
For Use With (Equipment) 3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer
Product Type Cathode Buffer Container
Shipping Condition Wet Ice
Content And Storage Each package contains 4 buffer containers.
Store at 2 - 8°C.
For In Vitro Diagnostic Use Only